3-(Methylamino)oxetane 98%

Supplier: Ambeed
Danger

Synonyms: N-Methyloxetan-3-amine

A290982-1G A290982-250MG A290982-5G
76816-580EA 146.41 USD
76816-580 76816-582 76816-584
3-(Methylamino)oxetane 98%
3-(Methylamino)oxetane

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.122 g/mol
MDL Number: MFCD08544349
CAS Number: 952182-03-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR