N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate
A290657-100G
A290657-1G
A290657-25G
A290657-5G
76816-572EA
412
USD
76816-572
76816-574
76816-576
76816-578
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10) 98%
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10)
Precaución: for Research Use Only
Formula:
C₁₅H₃₃NO₃S MW: 307.4924 g/mol |
MDL Number:
MFCD00036908 CAS Number: 15163-36-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...