(R)-(-)-Glycidyl butyrate 95%

Supplier: Ambeed
Warning

Synonyms: (-)-[(2R)-Oxiran-2-yl]methyl butanoate, (R)-Oxiran-2-ylmethyl butyrate

A290102-100G A290102-25G A290102-500G A290102-5G
76816-562EA 37.14 USD
76816-562 76816-564 76816-566 76816-568
(R)-(-)-Glycidyl butyrate 95%
(R)-(-)-Glycidyl butyrate

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD00075120
CAS Number: 60456-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR