2-(Aminomethyl)piperidine, 2-BOC protected 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-{[(tert-Butoxycarbonyl)amino]methyl}piperidine, tert-Butyl [(piperidin-2-yl)methyl]carbamate, tert-Butyl (piperidin-2-ylmethyl)carbamate
A289643-1G
A289643-250MG
A289643-25G
A289643-5G
76816-546EA
145.35
USD
76816-546
76816-548
76816-550
76816-552
76816-554
2-(Aminomethyl)piperidine, 2-BOC protected 97%
2-(Aminomethyl)piperidine, 2-BOC protected
Precaución: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ MW: 214.305 g/mol |
MDL Number:
MFCD02181068 CAS Number: 141774-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...