2-Methoxy-5-nitronicotinic acid 95%

Supplier: Ambeed
Warning

A289634-100MG A289634-1G A289634-250MG
76816-540EA 39.26 USD
76816-540 76816-542 76816-544
2-Methoxy-5-nitronicotinic acid 95%
2-Methoxy-5-nitronicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₅
MDL Number: MFCD11052413
CAS Number: 1020635-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR