2-Chloro-5-hydroxyisonicotinaldehyde 98%

Supplier: Ambeed
Warning

A288025-100MG A288025-250MG
76816-520EA 91.24 USD
76816-520 76816-522
2-Chloro-5-hydroxyisonicotinaldehyde 98%
2-Chloro-5-hydroxyisonicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MW: 157.554 g/mol
MDL Number: MFCD13189121
CAS Number: 1060804-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR