3-Methyl-1H-pyrazolo[3,4-c]pyridine 97%

Supplier: Ambeed
Warning

A287628-100MG A287628-1G A287628-250MG
76816-508EA 35 USD
76816-508 76816-510 76816-512
3-Methyl-1H-pyrazolo[3,4-c]pyridine 97%
3-Methyl-1H-pyrazolo[3,4-c]pyridine

Precaución: for Research Use Only

Formula: C7H7N3
MDL Number: MFCD13191779
CAS Number: 1072249-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR