(S)-5-Bromo-2,3-dihydro-1H-inden-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A286907-100MG A286907-1G A286907-250MG
76816-496EA 101.85 USD
76816-496 76816-498 76816-500
(S)-5-Bromo-2,3-dihydro-1H-inden-1-amine hydrochloride 95%
(S)-5-Bromo-2,3-dihydro-1H-inden-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MW: 248.54 g/mol
MDL Number: MFCD21603637
CAS Number: 916210-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR