Aminoacetamidine dihydrobromide 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminoacetamidine dihydrobromide

A286880-1G A286880-250MG A286880-5G
76816-490EA 47.75 USD
76816-490 76816-492 76816-494
Aminoacetamidine dihydrobromide 97%
Aminoacetamidine dihydrobromide

Precaución: for Research Use Only

Formula: C₂H₇N₃
MW: 234.92 g/mol
MDL Number: MFCD00671486
CAS Number: 69816-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR