2-Bromo-1,5-difluoro-3-nitrobenzene 95%

Supplier: Ambeed
Warning

A286875-1G A286875-250MG A286875-25G A286875-5G
76816-482EA 36.08 USD
76816-482 76816-484 76816-486 76816-488
2-Bromo-1,5-difluoro-3-nitrobenzene 95%
2-Bromo-1,5-difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MW: 237.986 g/mol
MDL Number: MFCD04112512
CAS Number: 877161-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR