(S)-Butyl-2-aminopropanoate 97%

Supplier: Ambeed
Warning

Synonyms: H-Ala-OBu, (S)-Butyl 2-aminopropanoate

A286528-250MG A286528-25G A286528-5G A286528-1G
76816-468EA 1357.96 USD
76816-468 76816-470 76816-472 76816-474 76816-476 77883-776
(S)-Butyl-2-aminopropanoate 97%
(S)-Butyl-2-aminopropanoate

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MDL Number: MFCD12796122
CAS Number: 2885-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR