1-tert-Butyl 3-ethyl azetidine-1,3-dicarboxylate 97%

Supplier: Ambeed
Warning

A286427-1G A286427-250MG A286427-25G A286427-5G
76816-454EA 29.71 USD
76816-454 76816-456 76816-458 76816-460
1-tert-Butyl 3-ethyl azetidine-1,3-dicarboxylate 97%
1-tert-Butyl 3-ethyl azetidine-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD08704361
CAS Number: 1346674-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR