(4-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride 98%

Supplier: Ambeed
Warning

A285832-250MG A285832-1G A285832-5G
76816-446EA 5 USD
76816-446 77612-646 77883-774
(4-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride 98%
(4-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₅BClNO₂
MDL Number: MFCD10698488
CAS Number: 938465-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR