4-[Bis(4-methoxyphenyl)amino]benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Bis(4-methoxyphenyl)amino)benzaldehyde, 4-Formyl-4',4''-dimethoxytriphenylamine

A285618-100MG A285618-250MG A285618-5G A285618-1G
76816-440EA 35.01 USD
76816-440 76816-442 76816-444 77612-644
4-[Bis(4-methoxyphenyl)amino]benzaldehyde 98%
4-[Bis(4-methoxyphenyl)amino]benzaldehyde

Precaución: for Research Use Only

Formula: C₂₁H₁₉NO₃
MW: 333.39 g/mol
MDL Number: MFCD16659083
CAS Number: 89115-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR