(S)-4,4-Difluoropyrrolidine-2-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A285491-100MG A285491-1G A285491-250MG
76816-436EA 17 USD
76816-436 76816-438 77883-773
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid hydrochloride 95%
(S)-4,4-Difluoropyrrolidine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇F₂NO₂·ClH
MDL Number: MFCD22199978
CAS Number: 1588480-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR