2-(3,4-Dihydroxyphenyl)acetonitrile 97%

Supplier: Ambeed
Warning

A284692-100MG A284692-250MG
76816-426EA 318.27 USD
76816-426 76816-428
2-(3,4-Dihydroxyphenyl)acetonitrile 97%
2-(3,4-Dihydroxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MW: 149.14 g/mol
MDL Number: MFCD00236011
CAS Number: 1126-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR