Daclatasvir dihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: BMS 790052-05, Daclatasvir 2HCl, Methyl ((S)-1-((S)-2-(5-(4'-(2-((S)-1-((methoxycarbonyl)-L-valyl)pyrrolidin-2-yl)-1H-imidazol-5-yl)-[1,1'-biphenyl]-4-yl)-1H-imidazol-2-yl)pyrrolidin-1-yl)-3-methyl-1-oxobutan-2-yl)carbamate dihydrochloride, Methyl N-[(2S)-1-[(2S)-2-[5-[4-[4-[2-[(2S)-1-[(2S)-2-(methoxycarbonylamino)-3-methylbutanoyl]pyrrolidin-2-yl]-1H-imidazol-5-yl]phenyl]phenyl]-1H-imidazol-2-yl]pyrrolidin-1-yl]-3-methyl-1-oxobutan-2-yl]carbamate dihydrochloride

A284426-10MG A284426-50MG
76816-422EA 5.31 USD
76816-422 76816-424
Daclatasvir dihydrochloride 98+%
Daclatasvir dihydrochloride

Precaución: for Research Use Only

Formula: C₄₀H₅₀N₈O₆·2HCl
MW: 811.8 g/mol
MDL Number: MFCD25541736
CAS Number: 1009119-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR