2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-oxo-7-azaspiro[3.5]nonane-7-carboxylate, 7-Azaspiro[3.5]nonan-2-one, N-BOC protected, 2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate tert-butyl ester
A284079-100G
A284079-10G
A284079-1G
A284079-250MG
A284079-5G
A284079-25G
A284079-500G
76816-412EA
267
USD
76816-412
76816-414
76816-416
76816-418
76816-420
77550-742
77672-466
2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester 97%
2-Oxo-7-azaspiro[3.5]nonane-7-carboxylate-tert-butyl ester
Precaución: for Research Use Only
Formula:
C₁₃H₂₁NO₃ MW: 239.32 g/mol |
MDL Number:
MFCD12198552 CAS Number: 203661-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...