(3-Bromo-5-ethoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-ethoxyphenylboronic acid

A283095-100MG A283095-1G A283095-250MG
76816-398EA 5 USD
76816-398 76816-400 76816-402
(3-Bromo-5-ethoxyphenyl)boronic acid 95%
(3-Bromo-5-ethoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₃
MDL Number: MFCD07369739
CAS Number: 849062-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR