(2-Fluoro-6-isopropoxyphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluro-6-isopropoxyphenylboronic acid

A282861-5G A282861-1G
76816-396EA 51.01 USD
76816-396 77672-462
(2-Fluoro-6-isopropoxyphenyl)boronic acid 95%
(2-Fluoro-6-isopropoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
MDL Number: MFCD06798078
CAS Number: 870777-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR