Methyl 2-(piperidin-1-ylmethyl)benzoate 98+%

Supplier: Ambeed
Warning

A280980-1G A280980-250MG A280980-5G
76816-386EA 65.78 USD
76816-386 76816-388 76816-390
Methyl 2-(piperidin-1-ylmethyl)benzoate 98+%
Methyl 2-(piperidin-1-ylmethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.30 g/mol
MDL Number: MFCD08272686
CAS Number: 914347-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR