tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Diaza-spiro[3.3]heptane-2-carboxylic acid tert-butyl ester oxalate salt, tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate

A280524-1G A280524-250MG A280524-25G A280524-5G
76816-378EA 103 USD
76816-378 76816-380 76816-382 76816-384
tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate 95%
tert-Butyl-2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₆
MW: 288.3 g/mol
MDL Number: MFCD11976177
CAS Number: 1227382-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR