4-(4-Chlorophenylcarbamoyl)phenylboronic acid 97+%

Supplier: Ambeed
Warning

Synonyms: (4-((4-Chlorophenyl)carbamoyl)phenyl)boronic acid

A280512-1G A280512-250MG A280512-5G
76816-372EA 11 USD
76816-372 76816-374 76816-376
4-(4-Chlorophenylcarbamoyl)phenylboronic acid 97+%
4-(4-Chlorophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BClNO₃
MDL Number: MFCD09027203
CAS Number: 874288-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR