2-Isoproxy-3-methylpyridine-5-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (6-Isopropoxy-5-methylpyridin-3-yl)boronic acid

A280403-1G A280403-250MG A280403-5G
76816-362EA 62.6 USD
76816-362 76816-364 76816-366
2-Isoproxy-3-methylpyridine-5-boronic acid 98%
2-Isoproxy-3-methylpyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₃
MW: 195.023 g/mol
MDL Number: MFCD13181629
CAS Number: 1402238-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR