1-Cyclohexyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A279990-100MG A279990-1G A279990-250MG A279990-5G
76816-350EA 36 USD
76816-350 76816-352 76816-354 76816-356
1-Cyclohexyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 95%
1-Cyclohexyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₅H₂₅BN₂O₂
MDL Number: MFCD16659787
CAS Number: 1175275-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR