6-Chlorobenzofuran-3(2H)-one 98%

Supplier: Ambeed
Warning

A279562-250MG A279562-5G
76816-348EA 19 USD
76816-348 77881-761
6-Chlorobenzofuran-3(2H)-one 98%
6-Chlorobenzofuran-3(2H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClO₂
MDL Number: MFCD06738725
CAS Number: 3260-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR