N,2,2-Trimethyl-1-propanamine hydrochloride 98%

Supplier: Ambeed
Warning

A279362-1G A279362-250MG
76816-344EA 133 USD
76816-344 76816-346
N,2,2-Trimethyl-1-propanamine hydrochloride 98%

Precaución: for Research Use Only

Formula: C₆H₁₆ClN
MDL Number: MFCD00154107
CAS Number: 31820-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR