2-Chloro-(5-ethoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-(ethoxycarbonyl)phenylboronic acid, (2-Chloro-5-(ethoxycarbonyl)phenyl)boronic acid

A278891-1G A278891-250MG A278891-25G A278891-5G
76816-336EA 103.97 USD
76816-336 76816-338 76816-340 76816-342
2-Chloro-(5-ethoxycarbonyl)phenylboronic acid 98%
2-Chloro-(5-ethoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BClO₄
MW: 228.4373 g/mol
MDL Number: MFCD08689529
CAS Number: 913835-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR