L-Asparagine methyl ester hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A276694-1G
A276694-250MG
A276694-5G
76816-316EA
32
USD
76816-316
76816-318
76816-320
L-Asparagine methyl ester hydrochloride 98%
L-Asparagine methyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₁ClN₂O₃ |
MDL Number:
MFCD01317794 CAS Number: 57461-34-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...