2,6-Dichloro-4-iodophenol 98%

Supplier: Ambeed
Warning

A275136-1G A275136-250MG A275136-5G
76816-296EA 10 USD
76816-296 76816-298 76816-300
2,6-Dichloro-4-iodophenol 98%
2,6-Dichloro-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃Cl₂IO
MDL Number: MFCD00466446
CAS Number: 34074-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR