(2-Formyl-4-methylphenyl)boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 2-Formyl-4-methylphenylboronic acid

A275054-1G A275054-250MG A275054-5G
76816-290EA 146 USD
76816-290 76816-292 76816-294
(2-Formyl-4-methylphenyl)boronic acid 95+%
(2-Formyl-4-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
MDL Number: MFCD13181634
CAS Number: 444188-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR