4-Bromo-2,5-difluorobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A274707-100G
A274707-10G
A274707-1G
A274707-250MG
A274707-25G
A274707-5G
76816-272EA
464.68
USD
76816-272
76816-274
76816-276
76816-278
76816-280
76816-282
4-Bromo-2,5-difluorobenzoic acid 97%
4-Bromo-2,5-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃BrF₂O₂ MW: 237.0000 g/mol |
MDL Number:
MFCD00844105 CAS Number: 28314-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...