2,4-Dimethyl-5-nitrobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A272489-10G
A272489-1G
A272489-25G
A272489-5G
76816-224EA
187.78
USD
76816-224
76816-226
76816-228
76816-230
2,4-Dimethyl-5-nitrobenzoic acid 98%
2,4-Dimethyl-5-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₉NO₄ MW: 195.17 g/mol |
MDL Number:
MFCD01463419 CAS Number: 220504-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...