3-Methoxy-2-nitroaniline 97%

Supplier: Ambeed
Warning

A272236-100MG A272236-1G A272236-250MG A272236-5G
76816-206EA 5.31 USD
76816-206 76816-208 76816-210 76816-212
3-Methoxy-2-nitroaniline 97%
3-Methoxy-2-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD18324417
CAS Number: 16554-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR