5-Chloro-2-ethynylpyridine 98%

Supplier: Ambeed
Warning

A271622-1G A271622-100MG A271622-5G
76816-200EA 52 USD
76816-200 77883-749 77883-750
5-Chloro-2-ethynylpyridine 98%
5-Chloro-2-ethynylpyridine

Precaución: for Research Use Only

Formula: C₇H₄ClN
MDL Number: MFCD13189715
CAS Number: 1196153-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR