1-Bromo-3,4,5-trifluoro-2-nitrobenzene 98%

Supplier: Ambeed
Danger

A271219-1G A271219-25G A271219-5G
76816-194EA 25.47 USD
76816-194 76816-196 76816-198
1-Bromo-3,4,5-trifluoro-2-nitrobenzene 98%
1-Bromo-3,4,5-trifluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆HBrF₃NO₂
MW: 255.977 g/mol
MDL Number: MFCD21603959
CAS Number: 1416373-06-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR