2-Bromo-5-(trifluoromethoxy)phenol 97%

Supplier: Ambeed
Warning

A270515-1G A270515-5G
76816-190EA 9 USD
76816-190 77883-745
2-Bromo-5-(trifluoromethoxy)phenol 97%
2-Bromo-5-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O₂
MDL Number: MFCD06660162
CAS Number: 205371-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR