2-Amino-6-chloronicotinonitrile 95%
Supplier: Ambeed
|
Warning
|
A270298-100MG
A270298-1G
A270298-250MG
A270298-5G
76816-182EA
75.33
USD
76816-182
76816-184
76816-186
76816-188
2-Amino-6-chloronicotinonitrile 95%
2-Amino-6-chloronicotinonitrile
Precaución: for Research Use Only
Formula:
C₆H₄ClN₃ MW: 153.569 g/mol |
MDL Number:
MFCD18261104 CAS Number: 52471-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...