5-Fluoro-2-methylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A269863-250MG A269863-1G A269863-100MG
76816-172EA 96.55 USD
76816-172 77612-582 77612-580
5-Fluoro-2-methylpyrimidin-4-amine 95%
5-Fluoro-2-methylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₅H₆FN₃
MW: 127.12 g/mol
MDL Number: MFCD23099431
CAS Number: 1416372-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR