H-D-Asp(OtBu)-OH 98%

Supplier: Ambeed
Warning

A269306-100G A269306-10G A269306-1G A269306-25G A269306-5G
76816-154EA 277 USD
76816-154 76816-156 76816-158 76816-160 76816-162
H-D-Asp(OtBu)-OH 98%
H-D-Asp(OtBu)-OH

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄
MDL Number: MFCD00077101
CAS Number: 64960-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR