(R)-Di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate 95%

Supplier: Ambeed
Warning

A267735-100MG A267735-1G A267735-250MG
76816-136EA 5.31 USD
76816-136 76816-138 76816-140
(R)-Di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate 95%
(R)-Di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₅
MW: 285.34 g/mol
MDL Number: MFCD16620747
CAS Number: 205524-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR