5-Methyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A267533-250MG A267533-5G
76816-130EA 26.53 USD
76816-130 76816-132
5-Methyl-1H-pyrazole-4-carboxylic acid 97%
5-Methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
MDL Number: MFCD08443868
CAS Number: 1035225-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR