trans,trans-4-n-Propyl-4'-[4-(trifluoromethoxy)phenyl]bicyclohexyl 97%

Supplier: Ambeed
Warning

A267458-10G A267458-1G A267458-250MG A267458-5G A267458-25G
76816-122EA 25 USD
76816-122 76816-124 76816-126 76816-128 77883-734
trans,trans-4-n-Propyl-4'-[4-(trifluoromethoxy)phenyl]bicyclohexyl 97%
trans,trans-4-n-Propyl-4'-[4-(trifluoromethoxy)phenyl]bicyclohexyl

Precaución: for Research Use Only

Formula: C₂₂H₃₁F₃O
MDL Number: MFCD20488041
CAS Number: 133937-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR