((4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)methyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

A266097-100G A266097-10G A266097-25G A266097-5G
76816-108EA 68 USD
76816-108 76816-110 76816-112 76816-114 76816-116
((4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)methyl)triphenylphosphonium bromide 97%
((4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)methyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₃₄H₃₄BrFN₃O₂PS
MDL Number: MFCD12911893
CAS Number: 885477-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR