L(+)-Asparagine monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: L-Asparagine monohydrate, H-Asn-OH·H₂O, (S)-(+)-Asparagine monohydrate, (S)-2,4-Diamino-4-oxobutanoic acid hydrate, H-Asn-OH.H2O

A265744-100G A265744-25G A265744-500G
76816-100EA 9 USD
76816-100 76816-102 76816-104
L(+)-Asparagine monohydrate 98%
L(+)-Asparagine monohydrate

Precaución: for Research Use Only

Formula: C₄H₈N₂O₃·1H₂O
MDL Number: MFCD00151038
CAS Number: 5794-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR