4-Chloro-1H-pyrrolo[2,3-b]pyridin-6-amine 95%

Supplier: Ambeed
Warning

A264948-100MG A264948-1G A264948-250MG A264948-5G
76816-078EA 42.44 USD
76816-078 76816-080 76816-082 76816-084
4-Chloro-1H-pyrrolo[2,3-b]pyridin-6-amine 95%
4-Chloro-1H-pyrrolo[2,3-b]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.60 g/mol
MDL Number: MFCD12924391
CAS Number: 935466-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR