Methyl-3-(cyanomethyl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(cyanomethyl)benzoate

A264478-100G A264478-500G A264478-5G
76816-060EA 58.35 USD
76816-060 76816-062 76816-064
Methyl-3-(cyanomethyl)benzoate 98%
Methyl-3-(cyanomethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD07783720
CAS Number: 68432-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR