1-Phenyl-1H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A263702-1G A263702-250MG A263702-5G
76816-050EA 69 USD
76816-050 76816-052 77672-360
1-Phenyl-1H-1,2,4-triazole 98%
1-Phenyl-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₈H₇N₃
MW: 145.161 g/mol
MDL Number: MFCD01076193
CAS Number: 13423-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR