Chroman-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A262822-100MG A262822-50MG A262822-250MG A262822-1G
76816-036EA 82 USD
76816-036 76816-038 77593-632 77883-726
Chroman-4-carboxylic acid 95%
Chroman-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD11848749
CAS Number: 20426-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR