(3-Chlorophenoxy)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-CPA, 3-Chlorophenoxyacetic acid

A261181-100G A261181-25G
76816-008EA 161.26 USD
76816-008 76816-010
(3-Chlorophenoxy)acetic acid 98%
(3-Chlorophenoxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
MDL Number: MFCD00015850
CAS Number: 588-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR